Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSLP Receptor Monoclonal Antibody (eBio1A6 (1A6)), eBioscience™
GREENER_CHOICE

Catalog No. 5013110
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-131-10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-131-10 Supplier Invitrogen™ Supplier No. 14549982
  • $309.00
    $231.65 / Each of 1

    Save $77.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The eBio1A6 monoclonal antibody reacts with human thymic stromal-derived lymphopoietin receptor (TSLPR). TSLPR is an approximately 50 kDa protein with significant similarity to the common gamma-chain. TSLPR complexes with IL-7R alpha (CD127) to form the high affinity receptor that binds thymic stromal-derived lymphopoietin (TSLP). Human TSLPR is expressed by monocytes and CD11c+ dendritic cells, and TSLP binding induces the expression of the Th2 cell-attracting chemokines CCL17 and CCL22. Furthermore, the TSLPR-induced activation of dendritic cells indirectly results in the increased secretion of Th cytokines IL-4, -5 and -13, which may be necessary for the regulation of CD4+ T cell homeostasis. In mice, deficiency of TSLPR has no effect on lymphocyte numbers, whereas double deficiency of TSLPR and common gamma-chain results in fewer lymphocytes than seen in mice deficient in the common gamma-chain alone. The eBio1A6 monoclonal antibody is able to cross-block binding of another anti-human TSLPR monoclonal antibody, 1D8. Applications Reported: This eBio1A6 (1A6) antibody has been reported for use in flow cytometric analysis, and immunoblotting (WB). Applications Tested: This eBio1A6 (1A6) antibody has been tested by flow cytometric analysis of human TSLPR-transfected Ba/F3 cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell num...

Thymic stromal lymphopoietin (TSLP) has recently been identified as an important factor capable of driving dendritic cell maturation and activation. It is involved in the positive selection of regulatory T cells, maintenance of peripheral CD4+ T cell homeostasis and the induction of CD4+ T cell-mediated allergic reaction. TSLP is also capable of supporting the growth of fetal liver and adult B cell progenitors and their differentiation to the IgM-positive stage of B cell development. Its receptor TSLP-R will bind TSLP in a low-affinity fashion in transfected cells; co-transfection with IL-7R-alpha cDNA results in high-affinity binding and a functional heteromeric complex. This heteromeric receptor requires stat5 for TSLP-mediated signal transduction and is inhibited by SOCS-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSLP Receptor
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone eBio1A6 (1A6)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CRLF2
Gene Accession No. Q9HC73
Gene Alias CRL2; Crlf2; CRLF2Y; Crlm2; CRLM-2; cytokine receptor CRL2 precusor; Cytokine receptor-like 2; Cytokine receptor-like factor 2; cytokine receptor-like molecule 2; delta 1; ILXR; IL-XR; Ly114; lymphocyte antigen 114; P2RY8/CRLF2 fusion; thymic stromal lymphopoietin protein receptor; thymic stromal-derived lymphopoietin receptor; thymic stromal-derived lymphopoietin, receptor; Tpte2; transmembrane phosphoinositide 3-phosphatase and tensin homolog 2; TSLP receptor; Tslpr; type I cytokine receptor delta 1; WI2-87105E3.1
Gene Symbols CRLF2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64109
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.