Learn More
Invitrogen™ TSLP Receptor Monoclonal Antibody (eBio1A6 (1A6)), Super Bright™ 600, eBioscience™

Description
The eBio1A6 monoclonal antibody reacts with human thymic stromal-derived lymphopoietin receptor (TSLPR). TSLPR is an approximately 50 kDa protein with significant similarity to the common gamma-chain. TSLPR complexes with IL-7R alpha (CD127) to form the high affinity receptor that binds thymic stromal-derived lymphopoietin (TSLP). Human TSLPR is expressed by monocytes and CD11c+ dendritic cells, and TSLP binding induces the expression of the Th2 cell-attracting chemokines CCL17 and CCL22. Furthermore, the TSLPR-induced activation of dendritic cells indirectly results in the increased secretion of Th cytokines IL-4, -5 and -13, which may be necessary for the regulation of CD4+ T cell homeostasis. In mice, deficiency of TSLPR has no effect on lymphocyte numbers, whereas double deficiency of TSLPR and common gamma-chain results in fewer lymphocytes than seen in mice deficient in the common gamma-chain alone. The eBio1A6 monoclonal antibody is able to cross-block binding of another anti-human TSLPR monoclonal antibody, 1D8. Applications Reported: This eBio1A6 (1A6) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio1A6 (1A6) antibody has been pre-titrated and tested by flow cytometric analysis of TSLPR transfected cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empiri...
Specifications
Specifications
| Antigen | TSLP Receptor |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | eBio1A6 (1A6) |
| Concentration | 5 μL/Test |
| Conjugate | Super Bright 600 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | CRLF2 |
| Gene Accession No. | Q9HC73 |
| Gene Alias | CRL2; Crlf2; CRLF2Y; Crlm2; CRLM-2; cytokine receptor CRL2 precusor; Cytokine receptor-like 2; Cytokine receptor-like factor 2; cytokine receptor-like molecule 2; delta 1; ILXR; IL-XR; Ly114; lymphocyte antigen 114; P2RY8/CRLF2 fusion; thymic stromal lymphopoietin protein receptor; thymic stromal-derived lymphopoietin receptor; thymic stromal-derived lymphopoietin, receptor; Tpte2; transmembrane phosphoinositide 3-phosphatase and tensin homolog 2; TSLP receptor; Tslpr; type I cytokine receptor delta 1; WI2-87105E3.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.