Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542948
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542948 Supplier Invitrogen™ Supplier No. PA542948
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LGILLPQFLG VLLTLLYITR VEDIIMEHSV TDGLLGPGAK PSVEAAGTGC Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

TSPAN15 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene. The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN15
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TSPAN15
Gene Accession No. O95858
Gene Alias 1110036D12Rik; 2700063A19Rik; AW048364; NET7; NET-7; Tetraspan NET-7; tetraspanin 15; tetraspanin-15; TM4SF15; transmembrane 4 superfamily member 15; transmembrane 4 superfamily member tetraspan NET-7; Tspan15; tspan-15; UNQ677/PRO1311
Gene Symbols TSPAN15
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TSPAN15 (aa 239-288).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23555
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.