Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592304
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592304 Supplier Invitrogen™ Supplier No. PA592304
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HDDLYDKKVD EEAEAKMLEK YKQERLEEMF PDEVDTPRDV AARIRFQKYR GLKSFRTSPW DPKENLPQDY ARIFQFQNFT NTRKSIFKEV EEKEVEGAEV GWYVTLHVSE VPVSVVECFR QGTPLIAFSL LPHEQKMSVL NMVVRRDPGN TEPVKAKEEL IFHCGFRRFR ASPLFSQHTA ADKHKLQRFL TADMALVATV YAPITFPPAS VLLFKQKSNG MHSLIATGHL MSVDPDRMVI KRVVLSGHPF KIFTKMAVVR YMFFNREDVL WFKPVELRTK WGRRGHIKEP LGTHGHMKCS FDGKLKSQDT VLMNLYKRVF PKWTYDPYVP EPVPWLKSEI SSTVPQGGME; Positive Samples: HepG2, Mouse testis; Cellular Location: Nucleus, nucleolus.

Required during maturation of the 40S ribosomal subunit in the nucleolus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSR1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TSR1
Gene Accession No. Q2NL82, Q5SWD9
Gene Alias AU040765; AW550801; KIAA1401; mKIAA1401; pre-rRNA-processing protein TSR1 homolog; Tsr1; TSR1 20S rRNA accumulation; TSR1, 20S rRNA accumulation, homolog; TSR1, 20S rRNA accumulation, homolog (S. cerevisiae); TSR1, ribosome maturation factor
Gene Symbols TSR1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 455-804 of human TSR1 (NP_0605983).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104662, 55720
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.