Learn More
Wanzl TSS Chrome Wire Basket
Description
- Add-on option for additional customization
- Offered in our Chrome Plus (CH) finish. The heavy nickel plate process features a hard chromium finish with special brighteners to enhance and increase the life of the shelving.
Specifications
Specifications
| Dimensions (L x W) | 18 x 48 in. |
| Includes | Wire Basket |
| Width (English) | 48 in. |
| For Use With (Application) | Shelving |
| Color | Chrome |
| Material | Steel |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.