Learn More
Wanzl TSS Clear Wire Shelf Label Holder
Description
- Add-on option for additional customization
- Clear
- Three ways to mount
Specifications
Specifications
| Color | Clear |
| For Use With (Application) | Shelving |
| Width (English) | 48 in. |
| Includes | Wire Shelf Label Holder |
| Material | Plastic |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.