Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSSK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110532
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110532 Supplier Invitrogen™ Supplier No. PA5110532
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEDFLLSNGY QLGKTIGEGT YSKVKEAFSK KHQRKVAIKV IDKMGGPEEF IQRFLPRELQ IVRTLDHKNI IQVYEMLESA DGKICLVMEL AEGGDVFDCV LNGGPLPESR AKALFRQMVE AIRYCHGCGV AHRDLKCENA LLQGFNLKLT DFGFAKVLPK SHRELSQTFC GSTAYAAPEV LQGIPHDSKK GDVWSMGVVL YVMLCASLPF DDTDIPKMLW QQQKGVSFPT HLSISADCQD LLKRLLEPDM ILRPSIEEVS WHPWLAST.

This gene encodes a kinase expressed exclusively in the testis that is thought to play a role in either germ cell differentiation or mature sperm function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSSK3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TSSK3
Gene Accession No. Q9D2E1
Gene Alias 1700014N07Rik; 4930594I21Rik; serine/threonine kinase 22C; serine/threonine kinase 22C (spermiogenesis associated); serine/threonine-protein kinase 22C; spermiogenesis associated; spermiogenesis associated 3; spermiogenesis associated serine/threonine kinase 22D; SPOGA3; STK22C; Stk22d; testis specific serine kinase 3; testis specific serine kinase-3; testis-specific kinase 3; testis-specific serine kinase 3; testis-specific serine/threonine kinase 22C; testis-specific serine/threonine kinase 3b; testis-specific serine/threonine-protein kinase 3; TSK3; TSK-3; Tssk3; Tssk-3; Tssk3b
Gene Symbols TSSK3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-268 of human TSSK3 (NP_443073.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297891, 58864
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.