Learn More
Medchemexpress LLC Tta-q6 | 910484-28-5 | 100.0% | 405.80 g·mol⁻¹ | C20H15ClF3N3O | 1 ML

Supplier: Medchemexpress LLC HY1038810MM 1ML
TTA-Q6 is a selective T-type calcium channel antagonist used for research into neurological diseases. It is provided as a powder and as a ready-to-use 10 mM solution in DMSO; molecular formula C20H15ClF3N3O, molecular weight 405.80 g·mol⁻¹.
- CAS number 910484-28-5.
- Molecular weight 405.80 g·mol⁻¹.
- Chemical formula C20H15ClF3N3O.
- Purity 99.97%.
- Available as solid (5-200 mg) and solution (10 mM, 1 mL).
- Soluble in DMSO 125 mg/mL; in vivo formulations ≥2.08 mg/mL in recommended vehicles.
- Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.