Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112983
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112983 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112983 Supplier Invitrogen™ Supplier No. PA5112983
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTC38 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC38
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TTC38
Gene Accession No. A3KMP2, Q5R3I4
Gene Alias 9030019H09; LL22NC03-5H6.5; RGD1306041; tetratricopeptide repeat domain 38; tetratricopeptide repeat protein 38; TPR repeat protein 38; TTC38
Gene Symbols TTC38
Host Species Rabbit
Immunogen Recombinant Human Tetratricopeptide repeat protein 38 protein (2-200AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239570, 55020
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.