Learn More
Biocare Medical, LLC TTF-1 Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC CM087C
Encompass_Preferred
Thyroid transcription factor-1 (TTF-1) is a member of the NKX2 family of homeodomain transcription factors. It is expressed in epithelial cells of the thyroid gland and the lung. Nuclei from liver, stomach, pancreas, small intestine, colon, kidney, breast, skin, testes, pituitary, prostate and adrenal glands are not reactive. TTF-1 is detected in primary lung adenocarcinomas and small cell carcinomas. It is absent in mesotheliomas, colon cancer and breast cancer. TTF-1 can be very useful when used in a panel with CK7, CK20, CDX2, Villin and CA125 antibodies.
Catalog No. 50-824-99
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.