Learn More
Diagnostic Biosystems TTF-1 Thyroid Transcription Factor-1, TTF-1, Thyroid and Lung Epithelial Marker
Supplier:  Diagnostic Biosystems PDM104
Encompass_Preferred
This antibody is specific to a 40 kDa protein, which is identified as thyroid transcription factor (TTF-1). TTF-1 is expressed in epithelial cells of the thyroid gland and lung. TTF-1 stains primary lung adenocarcinoma and small cell carcinoma but does not stain colon and breast carcinoma.
Catalog No. 50-238-5493
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.