Learn More
Invitrogen™ TTLL1 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, Molt-4, Raji, NIH-3T3. Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (85%), Xenopus laevis (91%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | TTLL1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | TTLL1 |
| Gene Accession No. | O95922, Q91V51 |
| Gene Alias | 6330444E16Rik; AV014541; bM121M7.1 (novel protein (ortholog of human tubulin tyrosine ligase-like 1 (TTLL1)), isoform 1); C22orf7; catalytic subunit of neural tubulin polyglutamylase; HS323M22B; p49; PGs3; Probable tubulin polyglutamylase TTLL1; TTLL1; Tubulin polyglutamylase complex subunit 3; tubulin tyrosine ligase like 1; tubulin tyrosine ligase-like 1; tubulin tyrosine ligase-like family member 1; tubulin tyrosine ligase-like family, member 1; tubulin-tyrosine ligase; tubulin--tyrosine ligase-like protein 1 |
| Gene Symbols | TTLL1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.