Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100050
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5100050 100 μL
1 options

Catalog No. PIPA5100050

Supplier: Invitrogen™ PA5100050

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TTLL6.

Predicted to enable tubulin binding activity and tubulin-glutamic acid ligase activity. Predicted to be involved in microtubule cytoskeleton organization and protein polyglutamylation. Predicted to act upstream of or within microtubule bundle formation; microtubule severing; and positive regulation of cilium movement. Located in ciliary basal body.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TTLL6
Gene Accession No. A4Q9E8, Q8N841
Gene Alias 4932418K24Rik; D11Moh43e; D11Moh44e; polyglutamylase; RGD1311922; t8130b59; TTL.6; TTLL6; Ttll6-ps1; Tubulin polyglutamylase TTLL6; tubulin tyrosine ligase like 6; tubulin tyrosine ligase-like family member 6; tubulin tyrosine ligase-like family, member 6; tubulin tyrosine ligase-like family, member 6, pseudogene 1; tubulin--tyrosine ligase-like protein 6
Gene Symbols TTLL6
Host Species Rabbit
Immunogen A synthesized peptide derived from human TTLL6(Accession Q8N841), corresponding to amino acid residues W91-P141.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237930, 284076, 287646
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.