Learn More
ProteinSimple iCE280/iCE3 FC Cartridge TTM Solution Kit
Supplier: ProteinSimple 102672
The TTM Solution Kit (2 bottles X 8 mL) is used for the Transfer Time Measurement (TTM) routine performed in Cartridge Installation and Start Up procedures with the FC-cIEF cartridge. This solution provides consistency.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.