Learn More
Hach Company Hach TU5400sc Ultra-High Precision Low-Range Laser Turbidimeter, EPA Version, System Check, RFID, Flow, 2-Channel
Supplier: Hach Company LXV525.97KTO038
The TU5400sc is an ultra-high precision, low-range online laser turbidimeter designed for continuous turbidity monitoring in water and wastewater applications. The EPA-version unit includes system check and RFID support, integrated flow measurement, and can be configured with an automatic cleaning module and controller interface for process control.
- Provides high precision low-range turbidity measurements with excellent repeatability.
- Compliant with EPA approved method for turbidity monitoring.
- Includes system check and RFID support for streamlined verification and calibration.
- Integrated flow measurement and optional automatic cleaning for continuous operation.
- Resolution of 0.0001 NTU and measurement range up to 700 NTU.
- Wall-mountable enclosure with IP55/IP65 protection for harsh environments.
- Operating range suitable for 0-50 °C and 5-95% relative humidity.