Learn More
Saint Gobain Performance Plastics TUB S-50-HL 5/8 x 7/8 50FT/PK
Supplier:  Saint Gobain Performance Plastics ADF00046
Encompass_Preferred
Saint-Gobain Tygon ND 100-65 Medical Tubing Diameter Inner 5/8 in. 15.9 mm Diameter Outer 7/8 in. 22.22 mm Overall Length 50 ft. 15.2 m Thickness 1/8 in. Non-DEHP
Catalog No. 50-206-8995
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.