Learn More
Saint Gobain Performance Plastics TUB SURG 1/4 x 3/8 50FT-15M/PK
Supplier:  Saint Gobain Performance Plastics ADF00017
Encompass_Preferred
Saint-Gobain Tygon ND 100-65 Medical Tubings Diameter Inner 1/4 in. Diameter Outer 3/8 in. Thickness 1/16 in. Non-DEHP Overall Length 50 ft. 15.2 m Clear
Catalog No. 50-206-8987
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.