Learn More
Saint Gobain Performance Plastics TUB TYG 2375 3/8 x 1/2 50FT/PK
Supplier:  Saint Gobain Performance Plastics AJK00027
Encompass_Preferred
Saint-Gobain Tygon Ultra-Chemical-Resistant Tubing Formula 2375 Diameter Inner 3/8 in. 9.5 mm Diameter Outer 1/2 in. 12.7 mm Thickness 1.6 mm 0.0625 in. Overall Length 50 ft. 15 m
Catalog No. 50-206-9027
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.