Learn More
Medchemexpress LLC HY-13271 10mg , Tubastatin A (Hydrochloride) CAS:1310693-92-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1327110MG
Encompass_Preferred
HY-13271 10mg Tubastatin A (Hydrochloride) CAS: 1310693-92-5 Tubastatin A Hydrochloride (Tubastatin A HCl) is a potent and selective HDAC6 inhibitor with IC50 of 15 nM in a cell-free assay, and is selective (1000-fold more) against all other isozymes except HDAC8 (57-fold more). Tubastatin A Hydrochloride also inhibits HDAC10 and metallo-β-lactamase domain-containing protein 2 (MBLAC2). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0732