Learn More
Tocris Bioscience™ Tubastatin A hydrochloride
Description
Specifications
Specifications
| Chemical Name or Material | N-Hydroxy-4-[(1,2,3,4-tetrahydro-2-methyl-5H-pyrido[4,3-b]indol-5-yl)methyl]benzamide hydrochloride |
| CAS | 1310693-92-5 |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Molecular Formula | C20H21N3O2.HCl |
| Formula Weight | Observed MW: 371.86 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.