Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUBB4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117495
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117495 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117495 Supplier Invitrogen™ Supplier No. PA5117495
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

TUBB4A (beta-4 tubulin) encodes a member of the beta tubulin family. Beta tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Mutations in this gene cause hypomyelinating leukodystrophy-6 and autosomal dominant torsion dystonia-4. Alternate splicing results in multiple transcript variants encoding different isoforms. A pseudogene of this gene is found on chromosome X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBB4A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TUBB4A
Gene Accession No. P04350, Q9D6F9
Gene Alias AI325297; beta-5; class IVa beta-tubulin; dystonia 4, torsion (autosomal dominant); DYT4; hypothetical protein LOC540236; M(beta)4; QtrA-11757; Tubb; TUBB4; Tubb4a; TUBB5; Tubulin 5 beta; tubulin beta; tubulin beta 4A class IVa; tubulin beta-4 chain; Tubulin beta-4A chain; tubulin, beta 4; tubulin, beta 4 class IVa; tubulin, beta 4A class IVa; tubulin, beta, 5; unnamed protein product
Gene Symbols TUBB4A
Host Species Rabbit
Immunogen Recombinant Human Tubulin beta-4A chain protein (1-444aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10382, 22153
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.