Learn More
Biotage TUBE 50ML GLASS W/1ML END PT
SDP
Supplier:  Biotage 030150
Encompass
50ml glass evaporation tube with 1 ml tip to fit in Horizon Technology's XcelVap stainless steel rack (pn:49541014). To be used with Horizon Technology's XcelVap nitrogen evaporation system (cat. #: NC0853177).
Catalog No. NC0926505
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.