Learn More
Globe Scientific™ Tube Adapters for Portable Clinical Centrifuge
Catalog No. 23556051
Change view
Click to view available options
Tube Size:
1.5/2 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tube Size
23-556-051 5 mL
23-556-052 1.5/2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-556-051 Supplier Globe Scientific™ Supplier No. GCCPADP5
Add to Cart
Add to Cart
This item is not returnable. View return policy
Tube adapters for the GCC-P Portable Clinical Centrifuge.
  • Ensures tubes are compatible with your Portable Clinical Centrifuge
Specifications
Type Tube Adapter
For Use With (Equipment) GCC-P Portable Clinical Centrifuge
Tube Size 5 mL
No. per Pack 24 Pk.