Learn More
Marketlab Tube Caps, 13 mm
Description
- Minimizes aerosol formation when opened
- Maintains a secure seal during centrifugation, agitation, refrigeration, and freezing
- Stretchable over-locking design helps reduce evaporation
- Can be applied and removed with one hand
Specifications
Specifications
| Product Type | Test Tube/Culture Tube Closure |
| Autoclavable | Not Autoclavable |
| Material | Polyethylene |
| Color | Lavender |
| Sterility | Non-sterile |
| Disposable | Single-use |