Learn More
Hach Company Centrifuge Tube, Conical Bottom, 10/pk
Supplier:  Hach Company 2115839
Encompass_Preferred
Centrifuge tube with conical bottom. Polycarbonate with molded-in 0.1mL graduations. For low-speed centrifugation up to 6000 x g. Autoclavable. The 15 mL size is also used with Settlometer Kit (P/N 21157-00). Supplied in packages of 10, without closures. Autoclavable. Nalge.
Catalog No. 50-199-5435
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.