Learn More
Research Products International Corp Tube Cube Test Tube Support System, Fluorescent Colors
SDP
Supplier:  Research Products International Corp 140442
Encompass_Preferred
Multi-option test tube support system accommodates a variety of tube sizes and styles. Each cube can be rotated independently within the tray for a particular tube size. Tube Cube modules and trays are constructed of durable polypropylene and can be autoclaved. Cube modules are color coded for convenience in sample identification.Each Tube Cube set consists of four removable modules and one storage tray.
Catalog No. 50-550-160
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.