Learn More
MilliporeSigma™ Supelco™ Tube Fitting, Cheminert™ 1/4-28 male UNF
Catalog No. 111011782 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
For Tubing 1/16 in. O.D.
For Tubing 1/8 in. O.D.
Diameter (Metric) Outer:
1.5 mm
3 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Diameter (Metric) Outer
11-101-1782 For Tubing 1/8 in. O.D. 3 mm
11-101-1869 For Tubing 1/16 in. O.D. 1.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-1782 Supplier MilliporeSigma™ Supelco™ Supplier No. 58712
Add to Cart
Add to Cart
Fittings and washers included. Chemically inert. Zero dead volume, small bore. Autoclavable. 500 psi (35 kg/cm3) pressure rating
TRUSTED_SUSTAINABILITY
Specifications
Color Mixed
Diameter (Metric) Outer 3 mm
Packaging Quantity 12/Pk.
Description For Tubing 1/8 in. O.D.
Material Stainless Steel
Cheminert is a registered trademark of Valco Instruments Co. Inc.