Learn More
MilliporeSigma™ Supelco™ Tube Fitting, Cheminert™ 1/4-28 male UNF (includes washer)
Catalog No. 111012000 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
10/Pk.
Catalog No. Packaging Quantity
11-101-2000 10/Pk.
1 options
Catalog No. 11-101-2000 Supplier MilliporeSigma™ Supelco™ Supplier No. 58711
Add to Cart
Add to Cart
Fittings and washers included. Chemically inert. Zero dead volume, small bore. Autoclavable. 500 psi (35 kg/cm3) pressure rating
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Diameter (Metric) Outer 3 mm
Packaging Quantity 10/Pk.
Description For Tubing 1/8 in. O.D.
Material Polypropylene
Cheminert is a registered trademark of Valco Instruments Co. Inc.