Learn More
Eppendorf™ Tube Holder 25/50 mL, for 4 x 25/50 mL conical tubes
Catalog No. 05414105 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
MixMate™
Catalog No. For Use With (Equipment)
05-414-105 MixMate™
1 options
Catalog No. 05-414-105 Supplier Eppendorf™ Supplier No. 5353040156
Add to Cart
Add to Cart
Mix your samples completely and reliably with the Eppendorf MixMate. 96 and 384-well plates and reaction tubes are mixed by the 2DMix-Control technology even at the microliter scale. The additional vortex function has a 2 s run timer allowing processing your next sample without restarting.
Tube Holder 25/50 mL, for 4 x 25/50 mL conical tubes- High-speed mixer (3,000 rpm) for fast and efficient mixing of small volumes in all common vessel formats up to 50 mL- 2DMix-Control technology (planar mixing orbit without vertical movement) for controlled mixing and reproducible results in seconds - Anti-spill technology for controlled mixing movements without lid wetting and cross-contamination- 5 different tube holders (0.5 mL, 1.5/2.0 mL, 5/15 mL, 30/50 mL, PCR 96-well plates) for high vessel flexibility- Integrated ergonomic vortex function for various vessel formats for distinct sample mixing- Minimal hand/arm vibration transfer during vortexing (certificate available) for ergonomic operation (PhysioCare Concept™)- 5 soft keys with pre-defined standard mixing parameters for easy usage- Very stable - remains stationary even at maximum mixing speed- Whisper quiet operation to benefit the work environment
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tube Holder
For Use With (Equipment) MixMate™