Learn More
MilliporeSigma™ Supelco™ Tube Holders
Catalog No. p-7203184 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
6 mm Sorbent Tubes
Detector Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-101-1633 Detector Tubes
11-101-1632 6 mm Sorbent Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-1633 Supplier MilliporeSigma™ Supelco™ Supplier No. 24869
Only null left
Add to Cart
Add to Cart
For use with PAS-500 Micro Air Sampler in Air Monitoring Applications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tube Holder
For Use With (Equipment) Detector Tubes
For Use With (Application) Air Sampling