Learn More
V&P Scientific Inc MAGNETIC RACK, For use with the VP 772F2M-1 Base, 12 Eppendorf or Microcentrifuge Tubes
Supplier:  V&P Scientific Inc VP 772F2R2
Encompass
MAGNETIC BEAD SEPARATION RACK, For use with the VP 772F2M-1 Base, 12 Eppendorf or Microcentrifuge Tubes, 5mL or 10mL or 15mL, 1 Sided Bead Pellet Location, Variable Positions, 48MGO, NdFeB Magnet, 23.5mm Center Spacing, 16.7mm Hole Diameter, Polycarbonate
Catalog No. NC2178665
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.