Learn More
Saint Gobain Performance Plastics Tube Tygon 1/4 x 7/16 50FT/PK
Supplier:  Saint Gobain Performance Plastics AAB00018
Encompass_Preferred
Saint-Gobain Tygon S3 Beverage Tubing Formula B-44-3 Overall Length 15.2 m 50 ft. Maximum Pressure 47 psi Diameter Inner 6.4 mm 1/4 in. Diameter Outer 11 mm 0.43 in. Autoclavable Autoclavable
Catalog No. 50-206-8914
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.