Learn More
Saint Gobain Performance Plastics Tube Tygon 3/16 x 3/8 50FT/PK
Supplier:  Saint Gobain Performance Plastics ACF00013
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Diameter Inner 3/16 in. Diameter Outer 3/8 in. Maximum Pressure 36 psi at 73 deg.F Thickness 3/32 in. Clear
Catalog No. 50-206-8943
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.