Learn More
Saint Gobain Performance Plastics Tube Tygon 3/16 x 5/16 50FT/PK
Supplier:  Saint Gobain Performance Plastics AAA00012
Encompass_Preferred
Saint-Gobain Tygon S3 Clear Tubing Formula B-44-4X Non-DEHP Diameter Inner 3/16 in. Diameter Outer 5/16 in. Maximum Pressure 9.5 MPa
Catalog No. 50-206-8905
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.