Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Tubed Media for Sartorius Membrane Filtration Units: Nutrient Broth

Catalog No. 5090117898 Shop All Sartorius Products
Change view
Click to view available options
Nutrient Broth, 50 x 20mL Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
5090117898 Nutrient Broth, 50 x 20mL Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5090117898 Supplier Sartorius Supplier No. 14132K

missing translation for 'validMainframeMsg'

Tubed media provides time and cost savings

The traditional culture media for microorganisms is agar media. This can be used for the membrane filtration method or for direct incubation. Agar media in tubes are for pouring agar plates. The content of one tube is sufficient for two 90mm or three 60mm Petri dishes.

  • 20mL tubes, 50 tubes per box
  • According to international formulas
  • Consistent quality

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Manufacturing Origin Germany
Media Type Nutrient Broth
Fill Volume 20 mL
For Use With (Application) Membrane filtration method or for direct incubation, for determination of total count
Packaging Type Bottle
Quantity 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.