Learn More
Hach Company Tubes, Micro Dist User Fill Option, Pkg of 10
Supplier:  Hach Company A17017
Encompass
Disposable Micro Dist distillation tubes (pkg of 10) designed for use with Micro Dist micro-distillation systems. These single-use tubes provide an alternative to traditional glassware, simplifying sample preparation and reducing the risk of cross-contamination during distillation of liquid or solid samples.
  • Disposable distillation tubes that replace glassware.
  • Accommodates liquid or solid samples.
  • Package contains 10 tubes.
  • Simplifies sample preparation and cleanup.
  • Reduces cross-contamination risk through single-use design.
Catalog No. NC2096516