Learn More
Beckman Coulter TUBES QS UC 1/2 X 2 (BOX-50)
Supplier:  Beckman Coulter 344075
Encompass_Preferred
Quick-Seal Ultra-Clear Tubes,Quantity:50,Size(mm):13 x 51, Vol.(mL): 5.1
Catalog No. NC9441337