Learn More
Description
- Compatible with 0.2 mL thermal cycler blocks
- Sold without caps
- Compatible with 0.2 mL thermal cycler blocks
- 8 tube strips
- Colored tubes are ideal for keeping track of samples
- Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays
- Applications PCR, qPCR/Real-Time PCR
Specifications
Specifications
| Certifications/Compliance | Certified DNA/RNase Free |
| Color | Red |
| Volume (Metric) Working | 0.2 mL |
| Volume (Metric) | 0.25 mL |
| Quantity | 250 strips |
| For Use With (Equipment) | Thermal Cyclers |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Reaction Speed | Standard |
| Closure Type | Not Included |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.