Learn More
California Advanced Labeling TubeWriter 360™ Direct High-Speed Labware Printing System
SDP
High-speed printing on any labware
Supplier:  California Advanced Labeling 36140100
Catalog No. 02-500-201
Description
Description
TubeWriter 360 ends manual labeling and allows mistake-free automated printing.
  • High-speed direct tube printing eliminates labels and messy handwriting
  • Prints on any labware — Microcentrifuge tubes, screw cap tubes, conical tubes, snap cap tubes, cluster tubes, cryogenic vials, histology cassettes, slides and microplates
  • Scannable barcode printing on demand (e.g., 2D Data Matrix, 39, 128, Interleaved 2 of 5)
  • Excel Interface or LIMS
  • Can print graphics, logos, and in other languages
Order Info
Installation and training is also available separately. (Contact Fisher Customer Service for details).
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.