Learn More
Tubewriter / California Advanced Labeling, Inc TubeWriter 400 - Replacement UV Curable Ink
SDP
Supplier:  Tubewriter / California Advanced Labeling, Inc TW4001
Encompass
Our patented UV-curable ink components allow for labeling directly onto curved plastics and glass surfaces, followed by UV curation to ensure printing stands up to harsh laboratory conditions.
Catalog No. NC2766451
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.