Learn More
SPEX CERTIPREP LLC 3P Turbidity QC Check Standards 5 NTU, 500 mL
Supplier:  SPEX CERTIPREP LLC 3P QCI156
Encompass_Preferred
(NSI by Zeptometrix) A formazin based CRM available as a 500mL bottle or as a case of 4 x 500mL bottles. Stable and ready to use as received. Do not freeze.
Catalog No. NC0828739