Learn More
MilliporeSigma™ Supelco™ Tubing Adapter 1/4-28 male UNF, male Luer
Catalog No. 111011636 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
11-101-1636 1/Pk.
1 options
Catalog No. 11-101-1636 Supplier MilliporeSigma™ Supelco™ Supplier No. 58722
Add to Cart
Add to Cart
Tubing adaptor is a national pipe thread taper made up of stainless steel which is used to connect a female adaptor. It may be used to connect capillary tubes in HPLC, GC etc.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Description for chromatography applications
Material CTFE
Valco is a registered trademark of Valco Instruments Co. Inc.