Learn More
MilliporeSigma™ Supelco™ Tubing Adapter 1/4-28 male UNF, for tube 1/4 in. O.D.
Catalog No. 111011872 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
11-101-1872 1/Pk.
1 options
Catalog No. 11-101-1872 Supplier MilliporeSigma™ Supelco™ Supplier No. 58742
Add to Cart
Add to Cart
Tubing adaptor is a national pipe thread taper made up of stainless steel which is used to connect a female adaptor. It may be used to connect capillary tubes in HPLC, GC etc.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Description For Tubing 1/4 in. O.D.
Material CTFE