Learn More
Cytiva TUBING CONNECTOR 1/8
Supplier:  Cytiva 18112117
Encompass
Connector (black PEEK) for use with tubing used on KTA systems. This 5/16 in connector works with OD 1/8 in tubing. Use with Ferrule for OD 1/8 in tubing. 10/pk.
Catalog No. 50-105-5658