Learn More
United Scientific™ Tubing Connectors Assortment
SDPSCI_ED
Catalog No. S15718 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Diameter (Metric) Inner:
Various
Catalog No. Diameter (Metric) Inner
S15718 Various
1 options
Catalog No. S15718 Supplier United Scientific™ Supplier No. TCA002
Add to Cart
Add to Cart
Provides a large assortment of tubing connectors.
  • A convenient assortment pack of 36 tubing connectors.
  • Compatible with a variety of tubing sizes and connections
  • Autoclavable
  • Pack includes:
    • 3 L-shaped connectors for 6 mm tubing
    • 3 L-shaped connectors for 8 mm tubing
    • 3 L-shaped connectors for 10 mm tubing
    • 3 T-shaped connectors for 6 mm tubing
    • 3 T-shaped connectors for 8 mm tubing
    • 3 T-shaped connectors for 10 mm tubing
    • 3 Y-shaped connectors for 6 mm tubing
    • 3 Y-shaped connectors for 8 mm tubing
    • 3 Y-shaped connectors for 10 mm tubing
    • 3 four-way connectors for 6 mm tubing
    • 3 four-way connectors for 8 mm tubing
    • 3 four-way connectors for 10 mm tubing
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner Various
Material Polypropylene
Diameter (English) Outer Various
Diameter (Metric) Outer Various
Overall Length (English) Various
Overall Length (Metric) Various
Color White
Product Type Connector