Learn More
AVANTOR MASTERFLEX (VWR LLC) Hand-Held Tubing Cutter, PTFE Coated Steel, Max 1-1/4"" OD Tubing
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0643890
Encompass
Hand-Held Tubing Cutter Steel blade coated with PTFE resin cuts through tubing up to 1 1/4 inch ID (cuts polyethylene tubing up to 1 inch ID)
Catalog No. NC9556527
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.