Learn More
IDEX Health & Science Upchurch TUBING PEEK 100' ROLL OF 1538
Supplier:  IDEX Health & Science Upchurch 1538XL
Encompass_Preferred
Idex PEEK Chromatography Tubing, Natural, Diameter Inner: 0.04 in., Overall Length: 100 ft., Pressure: 5000 psi, PEEK Tubing, For Use With: For High-Pressure Applications
Catalog No. NC1919825
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.