Learn More
MilliporeSigma™ Supelco™ Tubing Reamer
Catalog No. 111004943 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Tubing Reamer
Catalog No.
11-100-4943 Tubing Reamer
1 options
Catalog No. 11-100-4943 Supplier MilliporeSigma™ Supelco™ Supplier No. 20389
Add to Cart
Add to Cart
This 5 in. hand tool does a fine job of opening and rounding tubing ends thet are cut. The tip is stainless steel for durability and inertness; and the handle is wood for comfort.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Gas Chromatography (GC)