Learn More
Saint Gobain Performance Plastics TUBING VINCON PVC 100FT/PK
Supplier:  Saint Gobain Performance Plastics ABH02028
Encompass_Preferred
Saint-Gobain Vincon Flexible PVC Tubing Diameter Inner 3/8 in. 9.5 mm Diameter Outer 9/16 in. 14.3 mm Thickness 3/32 in. 2.4 mm Max. Operating Temperature 165 deg.F 74 deg.C Overall Length 100 ft. 30.5 m
Catalog No. 50-206-8933
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.