Learn More
Saint Gobain Performance Plastics TUBNG TYGN 1INID x 1.375INOD25FT
Supplier:  Saint Gobain Performance Plastics ACG42064
Encompass_Preferred
Saint-Gobain Tygon Formula 2475 High-Purity Tubing Diameter Inner 1 in. Diameter Outer 1.375 in. Thickness 4.8 mm 0.19 in. Certifications/Compliance Meets USP Class VI FDA and NSF specifications. Documented biocompatibility to ISO 10993 standards. Maximum Pressure 20 psi
Catalog No. 50-206-8982
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.